Learn More
ACROBiosystems Recombinant Protein;Human Carbonic Anhydrase IX / CA9 (138-414) Protein;HEK293;1MG;CA9,CAIX,CA9,CA-IX,G250,MN,Carbonic anhydrase 9,P54/58N,pMW1
Catalog No. 501658987
Encompass_Preferred
Recombinant Protein;Human Carbonic Anhydrase IX / CA9 (138-414) Protein;1MG;Human CA9, His Tag (CA9-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 138 - Asp 414 (Accession # NP_001207.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30.9 kDa. The protein migrates as 36 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8987 Supplier ACROBiosystems Supplier No. CA9H52201MG
Recombinant Protein;Human Carbonic Anhydrase IX / CA9 (138-414) Protein;1MG;Human CA9, His Tag (CA9-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 138 - Asp 414 (Accession # NP_001207.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30.9 kDa. The protein migrates as 36 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.