Learn More
Medchemexpress LLC Carbonyl cyanide (m-chlorophenyl)hydrazone | 555-60-2 | MFCD00001848 | >=97.0% | 204.61 g/mol | C9H5ClN4 | 250 MG
SDP
Catalog No. 5000212250
Encompass_Preferred
CCCP (carbonyl cyanide m-chlorophenylhydrazone) is a mitochondrial oxidative phosphorylation (OXPHOS) uncoupler used in research to depolarize mitochondrial membranes and disrupt cellular respiration. It is commonly applied to study mitophagy and mitochondrial quality-control pathways, for example inducing PINK1 activation and Parkin Ser65 phosphorylation.
  • Causes rapid mitochondrial membrane depolarization
  • Induces PINK1 activation and Parkin Ser65 phosphorylation for mitophagy studies
  • Useful to perturb cellular respiration and study reactive oxygen species production
  • Provided as a high-purity research reagent for laboratory experiments
Catalog No. 50-002-12250 Supplier Medchemexpress LLC Supplier No. HY100941250MG
CCCP (carbonyl cyanide m-chlorophenylhydrazone) is a mitochondrial oxidative phosphorylation (OXPHOS) uncoupler used in research to depolarize mitochondrial membranes and disrupt cellular respiration. It is commonly applied to study mitophagy and mitochondrial quality-control pathways, for example inducing PINK1 activation and Parkin Ser65 phosphorylation.
  • Causes rapid mitochondrial membrane depolarization
  • Induces PINK1 activation and Parkin Ser65 phosphorylation for mitophagy studies
  • Useful to perturb cellular respiration and study reactive oxygen species production
  • Provided as a high-purity research reagent for laboratory experiments