Learn More
Medchemexpress LLC Carbonyl cyanide m-chlorophenylhydrazone | 555-60-2 | MFCD00001848 | 99.8% | 204.62 g/mol | C9H5ClN4 | 500 MG
SDP
Catalog No. 5000212281
Encompass_Preferred
Carbonyl cyanide m-chlorophenylhydrazone (CCCP) is a mitochondrial oxidative phosphorylation (OXPHOS) uncoupler used to dissipate the mitochondrial membrane potential and probe mitochondrial function, mitophagy, and PINK1/Parkin signaling in cell biology research. It is provided as a solid or solution for preparation of DMSO stocks and experimental treatments.
  • Rapidly collapses mitochondrial membrane potential.
  • Activates PINK1 and promotes Parkin Ser65 phosphorylation.
  • Suitable for mitophagy and mitochondrial dynamics studies.
  • Soluble in DMSO for convenient stock solution preparation.
  • Supplied in multiple pack sizes to support varied experimental needs.
Catalog No. 50-002-12281 Supplier Medchemexpress LLC Supplier No. HY100941500MG
Carbonyl cyanide m-chlorophenylhydrazone (CCCP) is a mitochondrial oxidative phosphorylation (OXPHOS) uncoupler used to dissipate the mitochondrial membrane potential and probe mitochondrial function, mitophagy, and PINK1/Parkin signaling in cell biology research. It is provided as a solid or solution for preparation of DMSO stocks and experimental treatments.
  • Rapidly collapses mitochondrial membrane potential.
  • Activates PINK1 and promotes Parkin Ser65 phosphorylation.
  • Suitable for mitophagy and mitochondrial dynamics studies.
  • Soluble in DMSO for convenient stock solution preparation.
  • Supplied in multiple pack sizes to support varied experimental needs.