Learn More
Cusabio Technology LLC carbonyl reductase 1
Catalog No. 5000050021
Encompass_Preferred
The Human Carbonyl Reductase [NADPH] 1 (CBR1/CBR/CRN) ELISA Kit is designed to quantitatively measure CBR1 levels in serum plasma and tissue homogenates using a sandwich assay format This kit detects CBR1 within a broad dynamic range of 31 25 pg/mL to 2000 pg/mL with a sensitivity of 7 81 pg/mL ensuring precise quantification of low-abundance samples The assay requires a short incubation period of 1-5 hours and uses a sample volume of 50-100 L per test Detection is performed at 450 nm providing reliable and reproducible results for research applications Suitable for both routine laboratory use and specialized studies this kit offers a robust tool for analyzing CBR1 expression in diverse biological matrices
Catalog No. 50-000-50021 Supplier Cusabio Technology LLC Supplier No. CSBE17009H
The Human Carbonyl Reductase [NADPH] 1 (CBR1/CBR/CRN) ELISA Kit is designed to quantitatively measure CBR1 levels in serum plasma and tissue homogenates using a sandwich assay format This kit detects CBR1 within a broad dynamic range of 31 25 pg/mL to 2000 pg/mL with a sensitivity of 7 81 pg/mL ensuring precise quantification of low-abundance samples The assay requires a short incubation period of 1-5 hours and uses a sample volume of 50-100 L per test Detection is performed at 450 nm providing reliable and reproducible results for research applications Suitable for both routine laboratory use and specialized studies this kit offers a robust tool for analyzing CBR1 expression in diverse biological matrices