Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonylbis(triphenylphosphine)rhodium(I) Chloride, TCI America™
SDP

Catalog No. B16921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B16921G Supplier TCI America Supplier No. B16921G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13938-94-8
Molecular Formula C37H30ClOP2Rh
Molecular Weight (g/mol) 690.95
MDL Number MFCD00044273
InChI Key NGAGFUUIULZUPO-UHFFFAOYSA-M
Synonym Bis(triphenylphosphine)carbonylrhodium(I) Chloride, Carbonylchlorobis(triphenylphosphine)rhodium(I)
PubChem CID 131667324
IUPAC Name chlororhodiumylium; methanidylidyneoxidanium; bis(triphenylphosphane)
SMILES Cl[Rh+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 227°C
Quantity 1 g
Formula Weight 690.95
Physical Form Crystalline Powder
Chemical Name or Material Carbonylbis(triphenylphosphine)rhodium(I) Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.