Learn More
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Description
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II) is used as a raw material to synthesize other ru-hydride catalysts. It is involved in the hydrogenation of ring-opening metathesis polymers such as norbornene and other cyclic olefin ROMP polymers used as plastics, resins and optical materials. It can act as an alternative catalyst to rhodium hydrogenation catalyst. It also serves as a catalyst in the hydrogenation of conjugated diene copolymers like acrylonitrile-butadiene rubber (NBR).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 204°C to 206°C (decomposition) |
| Color | Gray |
| Linear Formula | [(C6H5)3P]3Ru(CO)(Cl)H |
| Quantity | 5 g |
| Sensitivity | Light sensitive; Very Hygroscopic |
| Solubility Information | Insoluble in water,ethanol,and DMSO. Sparingly soluble in benzene,chloroform and acetone. |
| Formula Weight | 952.41 |
| Physical Form | Crystalline |
| Chemical Name or Material | Carbonylchlorohydridotris (triphenylphosphine)ruthenium (II) |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 241-051-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.