Learn More
Thermo Scientific Dionex™ CarboPac™ PA20-Fast-4µm IC Column Microbore Separator
Catalog No. NC1698622 Shop All Thermo Scientific Products
Encompass
Type Microbore Separator | Flow Rate 0.25 mL/min. | pH 0 to 14 | Length (Metric) 100 mm | Particle Size 4 µm | Diameter (Metric) 2 mm | Max. Pressure 5000 psi (345 bar) | Temperature (Metric) 4°C to 60°C | For Use With (Application) Carbohydrate Analysis | Column Capacity 15 µeq | Column Construction PEEK | Column Chemistry Hydroxide | Mobile Phase Compatibility 0 to 50% Acetonitrile, Methanol, or Acetone | Anion Exchange Capacity 15 µeq |Resin Composition 4 µm Diameter Substrate (Ethylvinylbenzene 55% Crosslinked with Divinylbenzene) Agglomerated with 130 nm MicroBead Difunctional Quaternary Ammonium Ion (5.2% Crosslinked) | For Use With (Equipment) Dionex CarboPac PA20-Fast-4 µm Microbore Guard Column
Catalog No. NC1698622 Supplier Thermo Scientific Supplier No. 302749
Type Microbore Separator | Flow Rate 0.25 mL/min. | pH 0 to 14 | Length (Metric) 100 mm | Particle Size 4 µm | Diameter (Metric) 2 mm | Max. Pressure 5000 psi (345 bar) | Temperature (Metric) 4°C to 60°C | For Use With (Application) Carbohydrate Analysis | Column Capacity 15 µeq | Column Construction PEEK | Column Chemistry Hydroxide | Mobile Phase Compatibility 0 to 50% Acetonitrile, Methanol, or Acetone | Anion Exchange Capacity 15 µeq |Resin Composition 4 µm Diameter Substrate (Ethylvinylbenzene 55% Crosslinked with Divinylbenzene) Agglomerated with 130 nm MicroBead Difunctional Quaternary Ammonium Ion (5.2% Crosslinked) | For Use With (Equipment) Dionex CarboPac PA20-Fast-4 µm Microbore Guard Column