Learn More
Thermo Scientific Dionex™ CarboPac™ SA10-4µm IC Column, Microbore Separator
Catalog No. NC1619251 Shop All Thermo Scientific Products
Encompass
Separate simple sugars in biofuels and food samples quickly and with better resolution and easier peak identification and integration. The Thermo Scientific™ Dionex™ CarboPac™ SA10-4µm Column produces 60 to 80% higher peak efficiencies than the standard Dionex CarboPac SA10 columns, thus providing better resolution and easier peak identification and integration of these carbohydrates. The 4µm column analyzes six common sugars in food samples in only 10 minutes, or eight common biofuel mono- and disaccharides in less than 8 minutes.
Catalog No. NC1619251 Supplier Thermo Scientific Supplier No. 088235
Separate simple sugars in biofuels and food samples quickly and with better resolution and easier peak identification and integration. The Thermo Scientific™ Dionex™ CarboPac™ SA10-4µm Column produces 60 to 80% higher peak efficiencies than the standard Dionex CarboPac SA10 columns, thus providing better resolution and easier peak identification and integration of these carbohydrates. The 4µm column analyzes six common sugars in food samples in only 10 minutes, or eight common biofuel mono- and disaccharides in less than 8 minutes.