Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Carbopack™ Adsorbent, Matrix Carbopack™ X, 40-60 mesh

Catalog No. 111008503 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-8503 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-8503 Supplier MilliporeSigma™ Supelco™ Supplier No. 10436
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carbotrap™™/Carbopack™™ Adsorbent can withstand high bake-out temperature without losing the mechanical and chemical integrity.

Carbotrap™™/Carbopack™™ Adsorbent has a high affinity for isoprene and does not retain significant amounts of water. Furthermore it has a tendency to pulverize if it is packed too tightly.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 40/60 Mesh
Packaging Bottle
Quantity 10 g
Surface Area (Metric) ˜240 m2/g
Product Line Carbopack
Density ˜0.44 g/mL (free fall denstiy)

Carbopack is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.