Learn More
Medchemexpress LLC HY-17393 500mg , Carboplatin cis-Diammine(1,1-cyclobutanedicarboxylato)platinum CAS:41575-94-4 Purity:98%
SDP
Catalog No. 501492343
Encompass_Preferred
Medchemexpress, HY-17393 500mg Carboplatin cis-Diammine(1,1-cyclobutanedicarboxylato)platinum CAS:41575-94-4 Formula:C6H12N2O4Pt DNA Alkylator Purity:98% Carboplatin is an antitumor agent, with DNA-binding activity, interfering with duplication of DNA. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2343 Supplier Medchemexpress LLC Supplier No. HY17393500MG
Medchemexpress, HY-17393 500mg Carboplatin cis-Diammine(1,1-cyclobutanedicarboxylato)platinum CAS:41575-94-4 Formula:C6H12N2O4Pt DNA Alkylator Purity:98% Carboplatin is an antitumor agent, with DNA-binding activity, interfering with duplication of DNA. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.