Learn More
Medchemexpress LLC Carboplatin | 41575-94-4 | 99.9% | 371.25 | 200 MG
SDP
Catalog No. 5000384267
Encompass_Preferred
Carboplatin (NSC 241240) is a DNA synthesis inhibitor that works by binding to DNA, which in turn inhibits replication and transcription, ultimately inducing cell death. It is a derivative of CDDP and is considered a potent anti-cancer agent.
  • DNA synthesis inhibitor
  • Binds to DNA
  • Inhibits replication and transcription
  • Induces cell death
  • Derivative of CDDP
  • Potent anti-cancer agent
Catalog No. 50-003-84267 Supplier Medchemexpress LLC Supplier No. HY17393200MG
Carboplatin (NSC 241240) is a DNA synthesis inhibitor that works by binding to DNA, which in turn inhibits replication and transcription, ultimately inducing cell death. It is a derivative of CDDP and is considered a potent anti-cancer agent.
  • DNA synthesis inhibitor
  • Binds to DNA
  • Inhibits replication and transcription
  • Induces cell death
  • Derivative of CDDP
  • Potent anti-cancer agent