Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carboplatin | 41575-94-4 | MFCD00070464 | 99.98% | 371.25 | C6H12N2O4Pt | 10 MG
SDP

Catalog No. 5000298751
Encompass_Preferred

Carboplatin analytical standard supplied as a small, high-purity reference material for research and analytical applications. This material is intended for method development, calibration, and identity confirmation in laboratory settings and is not for human or clinical use.

  • High purity for analytical applications (≈99.98%).
  • Provided in a small, weighed package suitable for calibration and standards.
  • Includes chemical identifiers for traceability (CAS 41575-94-4).
  • Molecular formula C6H12N2O4Pt, molecular weight 371.25.
  • Intended for research use only; not for human administration.

Catalog No. 50-002-98751 Supplier Medchemexpress LLC Supplier No. HY17393R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carboplatin analytical standard supplied as a small, high-purity reference material for research and analytical applications. This material is intended for method development, calibration, and identity confirmation in laboratory settings and is not for human or clinical use.

  • High purity for analytical applications (≈99.98%).
  • Provided in a small, weighed package suitable for calibration and standards.
  • Includes chemical identifiers for traceability (CAS 41575-94-4).
  • Molecular formula C6H12N2O4Pt, molecular weight 371.25.
  • Intended for research use only; not for human administration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.