Learn More
Cayman Chemical CarboplatIn 50mg
SDP
Catalog No. 502954955
Encompass_Preferred
A DNA-crosslinking agent; induces the formation of DNA interstrand crosslinks and DNA-protein crosslinks at 200 and 300 µM; inhibits proliferation of GLC-4, A2780, and SKOV3 cells (IC50s = 22.7, 0.5, and 10.2 µM, respectively); reduces tumor growth in an ADJ/PC6 murine plasmacytoma model (ED90 = 0.6 mg/kg, i.p.)
Catalog No. 50-295-4955 Supplier Cayman Chemical Supplier No. 1311250 MG
A DNA-crosslinking agent; induces the formation of DNA interstrand crosslinks and DNA-protein crosslinks at 200 and 300 µM; inhibits proliferation of GLC-4, A2780, and SKOV3 cells (IC50s = 22.7, 0.5, and 10.2 µM, respectively); reduces tumor growth in an ADJ/PC6 murine plasmacytoma model (ED90 = 0.6 mg/kg, i.p.)