Learn More
Medchemexpress LLC Carboplatin-d4 | 41575-94-4 | 99.0% | 375.28 | 1 MG
SDP
Catalog No. 5000384580
Encompass_Preferred
Carboplatin-d4 is the deuterium labeled Carboplatin. Carboplatin (NSC 241240) is a DNA synthesis inhibitor that binds to DNA, inhibits replication and transcription, and induces cell death. It is a derivative of CDDP and a potent anti-cancer agent. Deuterium substitution can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-84580 Supplier Medchemexpress LLC Supplier No. HY17393S1MG
Carboplatin-d4 is the deuterium labeled Carboplatin. Carboplatin (NSC 241240) is a DNA synthesis inhibitor that binds to DNA, inhibits replication and transcription, and induces cell death. It is a derivative of CDDP and a potent anti-cancer agent. Deuterium substitution can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.