Learn More
Selleck Chemical LLC Carboplatin 200mg 41575-94-4 JM-8, CBDCA, NSC 241240
SDP
Catalog No. 507461
Encompass_Preferred
Carboplatin (JM-8, CBDCA, NSC 241240) is a DNA synthesis inhibitor by binding to DNA and interfering with the cell's repair mechanism in A2780, SKOV-3, IGROV-1, and HX62 cells. Solutions are best fresh-prepared. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-746-1 Supplier Selleck Chemical LLC Supplier No. S1215200MG
Carboplatin (JM-8, CBDCA, NSC 241240) is a DNA synthesis inhibitor by binding to DNA and interfering with the cell's repair mechanism in A2780, SKOV-3, IGROV-1, and HX62 cells. Solutions are best fresh-prepared. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.