Learn More
Crescent Chemical Co Inc CARBOPOL 934 PRACT.
SDP
Catalog No. 50247399
Encompass_Preferred
Carbopol 934 pract. A carboxyvinyl-polymer of Goodrich Chemical Co. with a very high molecular weight. The powder supplied as the free acid is soluble in water up to 5 % and forms a solution of fairly low viscosity with pH ca. 3. Upon neutralization with sodium h
Catalog No. 50-247-399 Supplier Crescent Chemical Co Inc Supplier No. SE15885.01
Carbopol 934 pract. A carboxyvinyl-polymer of Goodrich Chemical Co. with a very high molecular weight. The powder supplied as the free acid is soluble in water up to 5 % and forms a solution of fairly low viscosity with pH ca. 3. Upon neutralization with sodium h