Learn More
Medchemexpress LLC Carboprost tromethamine | 58551-69-2 | 99.9% | C25H47NO8 | 25 MG
SDP
Catalog No. 5000359164
Encompass_Preferred
Carboprost tromethamine is the synthetic 15-methyl analogue of prostaglandin F2α. It effectively promotes uterine contraction and significantly reduces bleeding during and after delivery. It is for research use only and not sold to patients. It has been reported to be 84-96% successful in treating persistent hemorrhage due to uterine atony.
  • Synthetic 15-methyl analogue of prostaglandin F2α
  • Effectively promotes uterine contraction
  • Significantly reduces bleeding during and after delivery
  • Intended for research purposes only
  • 84-96% successful in treating persistent hemorrhage due to uterine atony
Catalog No. 50-003-59164 Supplier Medchemexpress LLC Supplier No. 25MGHYA0195
Carboprost tromethamine is the synthetic 15-methyl analogue of prostaglandin F2α. It effectively promotes uterine contraction and significantly reduces bleeding during and after delivery. It is for research use only and not sold to patients. It has been reported to be 84-96% successful in treating persistent hemorrhage due to uterine atony.
  • Synthetic 15-methyl analogue of prostaglandin F2α
  • Effectively promotes uterine contraction
  • Significantly reduces bleeding during and after delivery
  • Intended for research purposes only
  • 84-96% successful in treating persistent hemorrhage due to uterine atony