Learn More
Chrom Tech Inc CarboSep CHO-620 Column, 6.5x300mm
SDP
Catalog No. NC1831031
Encompass
Calcium form ligand-exchange column. Ideal for the separation of monosaccharides and sugar alcohols.
Catalog No. NC1831031 Supplier Chrom Tech Inc Supplier No. CHO999753
Calcium form ligand-exchange column. Ideal for the separation of monosaccharides and sugar alcohols.