Learn More
Cayman Chemical Carbostyril 124, Purity: ≥95%, CAS: 19840-99-4, 100 mg
SDP
Catalog No. 502911345
Encompass_Preferred
A chromphore; has been used as a building block in the synthesis of luminescent lanthanide complexes
Catalog No. 50-291-1345 Supplier Cayman Chemical Supplier No. 34967100 MG
A chromphore; has been used as a building block in the synthesis of luminescent lanthanide complexes