Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Carbowax Polyethylene Glycol PEG 3350 Flakes, 54 to 58°C Melting range 1 KG
SDP

Catalog No. 5024915
Encompass_Preferred

CAS #25322-68-3 H(OCH₂CH₂)nOH F.W. 190 - 210

An embedding medium for microscopy and histochemistry. Gao, K.X., (1993). Polyethylene Glycol as an Embedment for Microscopy and Histochemistry. CRC Press; ISBN# 0-8493-4323-2.
Melting range: 54 to 58°C
Molecular weight: 3000-3700

Catalog No. 50-249-15 Supplier Electron Microscopy Sciences Supplier No. 19760
May include imposed supplier surcharges.
Add to Cart
Add to Cart

CAS #25322-68-3 H(OCH₂CH₂)nOH F.W. 190 - 210

An embedding medium for microscopy and histochemistry. Gao, K.X., (1993). Polyethylene Glycol as an Embedment for Microscopy and Histochemistry. CRC Press; ISBN# 0-8493-4323-2.
Melting range: 54 to 58°C
Molecular weight: 3000-3700

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.