Learn More
Electron Microscopy Sciences Carbowax Polyethylene Glycol, PEG 400 viscous liquid, 25322-68-3, 450ml
SDP
Catalog No. 5024918
Encompass_Preferred
Molecular Formula H(OCH2CH2)nOH, Carbowax Polyethylene Glycol is an embedding medium for microscopy and histochemistry.
Catalog No. 50-249-18 Supplier Electron Microscopy Sciences Supplier No. 19720
Molecular Formula H(OCH2CH2)nOH, Carbowax Polyethylene Glycol is an embedding medium for microscopy and histochemistry.