Learn More
Bioworld Carboxine, 250 mg
Catalog No. 50255712
Encompass_Preferred
Carboxin Fungicide that inhibits the oxidation of succinate in sensitive yeast and fungi.For in vitro lab use only. Not for human use or consumption.Synonym: CarbathiineAppearance: SolidDensity : 1.45IUPAC_Name: 6-methyl-N-phenyl-2,3-dihydro-1,4-oxathiine-5-carboxamideMelting Point: 91.1-91.7 ºCRTECS Number: RP4550000
Catalog No. 50-255-712 Supplier Bioworld Supplier No. 403100041
Carboxin Fungicide that inhibits the oxidation of succinate in sensitive yeast and fungi.For in vitro lab use only. Not for human use or consumption.Synonym: CarbathiineAppearance: SolidDensity : 1.45IUPAC_Name: 6-methyl-N-phenyl-2,3-dihydro-1,4-oxathiine-5-carboxamideMelting Point: 91.1-91.7 ºCRTECS Number: RP4550000