Learn More
AdipoGen Carboxy-DCF
SDP
Supplier:  AdipoGen CDXC0043M200
Encompass_Preferred
Chemical. CAS 111843-78-8. Formula C21H10Cl2O7. MW 445.21. Synthetic. Fluorescent intracellular pH indicator in the pH 4-5 range.
Catalog No. 50-256-1520