Learn More
AdipoGen Carboxy-DCFDA
SDP
Catalog No. 502561522
Encompass_Preferred
Chemical. CAS 127770-45-0. Formula C25H14Cl2O9. MW 529.28. Synthetic. Intracellular esterase substrate with low sensitivity of fluorescence to intracellar pH.
Catalog No. 50-256-1522 Supplier AdipoGen Supplier No. CDXC0042M100
Chemical. CAS 127770-45-0. Formula C25H14Cl2O9. MW 529.28. Synthetic. Intracellular esterase substrate with low sensitivity of fluorescence to intracellar pH.