Learn More
AdipoGen Carboxy-DCFDA N-SE
SDP
Supplier:  AdipoGen CDXC0059M050
Encompass_Preferred
Chemical. CAS 147265-60-9. Formula C29H17Cl2NO11. MW 626.35. Synthetic. 5-and-6-Carboxy-2,7-dichlorofluorescein diacetate, succinimidyl ester is a useful fluorescent tracer that can passively diffuse into cells and covalently label intracellular proteins, resulting in long-term cell labeling. The reagent itself is colorless and nonfluorescent but becomes brightly green fluorescent once it is hydrolyzed by intracellular esterases.
Catalog No. 50-256-1498