Learn More
Abcam DCFA (5(6)-Carbo x y-2',7'-dichlorofluorescein), green fluorophore, 50MG
Catalog No. 50996231
Encompass_Preferred
MW 445.21 g/mol, Purity >95%. Green fluorophore. Intracellular pH indicator in the pH 4-5 range. DFCA DA (ab145415) precursor. 2,7-Dichlorofluorescein (ab146332) derivative.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-231 Supplier Abcam Supplier No. AB14543950MG
MW 445.21 g/mol, Purity >95%. Green fluorophore. Intracellular pH indicator in the pH 4-5 range. DFCA DA (ab145415) precursor. 2,7-Dichlorofluorescein (ab146332) derivative.
The product is subject to the following: Abcam Restricted Use Statement