Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC γ-carboxy-DL-glutamic acid | 56271-99-9 | MFCD00065488 | 98.0% | 191.14 | C6H9NO6 | 5 MG
SDP

Catalog No. 5000283790
Encompass_Preferred

γ-Carboxy-DL-glutamic acid is a glutamic acid derivative supplied as a solid reagent for laboratory research. It is provided as the optically active DL form with defined purity and storage instructions to support stability during handling and short-term storage.

  • Purity: 98.0%.
  • Molecular formula: C6H9NO6.
  • Molecular weight: 191.14.
  • Appearance: white to off-white solid.
  • Packaging options: small research quantities (e.g., milligram scale).
  • Storage: sealed, away from moisture; powder: -80°C (2 years) or -20°C (1 year).
  • Intended use: research reagent only.

Catalog No. 50-002-83790 Supplier Medchemexpress LLC Supplier No. HYW1419555MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

γ-Carboxy-DL-glutamic acid is a glutamic acid derivative supplied as a solid reagent for laboratory research. It is provided as the optically active DL form with defined purity and storage instructions to support stability during handling and short-term storage.

  • Purity: 98.0%.
  • Molecular formula: C6H9NO6.
  • Molecular weight: 191.14.
  • Appearance: white to off-white solid.
  • Packaging options: small research quantities (e.g., milligram scale).
  • Storage: sealed, away from moisture; powder: -80°C (2 years) or -20°C (1 year).
  • Intended use: research reagent only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.