Learn More
Medchemexpress LLC Carboxy-PEG4-phosphonic acid | 1623791-69-4 | 96.0% | C11H23O9P | 50 MG
SDP
Catalog No. 5000353858
Encompass_Preferred
Carboxy-PEG4-phosphonic acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-53858 Supplier Medchemexpress LLC Supplier No. HY14130250MG
Carboxy-PEG4-phosphonic acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only