Learn More
Enzo Life Sciences Carboxy-PTIO, (10mg), CAS Number: 148819-94-7
SDP
Supplier:  Enzo Life Sciences ALX430001M010
Encompass_Preferred
Nitric oxide (NO) scavenger. Water soluble and stable nitric oxide radical scavenger that shows in chemical and biological systems antagonistic action against the free nitric oxide radical (NO·). The inhibitory effects were almost 2-fold stronger than those of NMMA (Prod. No. ALX-106-001) and NOARG (Prod. No. ALX-105-001). Reacts with nitric oxide in a stoichometric manner. Can be used for an improved detection of nitric oxide in activated macrophage cultures in combination with the Griess reagent (Prod. No. ALX-400-004). Protects against endotoxin shock. Alternative Name: 2-(4-Carboxyphenyl)-4,4,5,5-tetramethylimidazoline-1-oxyl-3-oxide . potassium salt. Formula: C14H16N2O4 . K. MW: 315.4. CAS Number: 148819-94-7. Purity: ≥99% (HPLC). Appearance: Blue solid. Solubility: Soluble in 100% ethanol, methanol, DMSO or dimethyl formamide. Highly soluble in water or aqueous buffers, e. g. PBS, pH 7.2. Long Term Storage: -20°C
Catalog No. NC1278521