Learn More
Medchemexpress LLC Carboxy-PTIO potassium | 148819-94-7 | 99.0% | 315.39 | 100 MG
SDP
Catalog No. 5000385695
Encompass_Preferred
Carboxy-PTIO potassium is a potent nitric oxide (NO) scavenger that reacts quickly with NO to produce NO2. It can prevent hypotension and endotoxic shock by directly scavenging NO in a lipopolysaccharide-stimulated rat model.
  • Potent nitric oxide (NO) scavenger.
  • Reacts quickly with NO to produce NO2.
  • Can prevent hypotension and endotoxic shock through direct NO scavenging.
Catalog No. 50-003-85695 Supplier Medchemexpress LLC Supplier No. HY18734A100MG
Carboxy-PTIO potassium is a potent nitric oxide (NO) scavenger that reacts quickly with NO to produce NO2. It can prevent hypotension and endotoxic shock by directly scavenging NO in a lipopolysaccharide-stimulated rat model.
  • Potent nitric oxide (NO) scavenger.
  • Reacts quickly with NO to produce NO2.
  • Can prevent hypotension and endotoxic shock through direct NO scavenging.