Learn More
Medchemexpress LLC Carboxy-PTIO potassium | 148819-94-7 | 99.0% | 315.39 | 25 MG
SDP
Catalog No. 5000384990
Encompass_Preferred
Carboxy-PTIO potassium is a potent nitric oxide (NO) scavenger that reacts quickly with NO to produce NO2. It can prevent hypotension and endotoxic shock by directly scavenging NO in a lipopolysaccharide-stimulated rat model. It is for research use only and not sold to patients.
  • Potent nitric oxide (NO) scavenger.
  • Can prevent hypotension and endotoxic shock.
  • In vitro, it significantly suppresses the stimulation of NO expression induced by Physalin A treatment and reverses decreased mTOR and p-mTOR levels with concomitant suppression of LC3 I to LC3 II conversions in A375-S2 cells.
  • In vivo, it improves hypotension, renal dysfunction, and survival rate in LPS-treated rats.
Catalog No. 50-003-84990 Supplier Medchemexpress LLC Supplier No. 25MGHY18734A
Carboxy-PTIO potassium is a potent nitric oxide (NO) scavenger that reacts quickly with NO to produce NO2. It can prevent hypotension and endotoxic shock by directly scavenging NO in a lipopolysaccharide-stimulated rat model. It is for research use only and not sold to patients.
  • Potent nitric oxide (NO) scavenger.
  • Can prevent hypotension and endotoxic shock.
  • In vitro, it significantly suppresses the stimulation of NO expression induced by Physalin A treatment and reverses decreased mTOR and p-mTOR levels with concomitant suppression of LC3 I to LC3 II conversions in A375-S2 cells.
  • In vivo, it improves hypotension, renal dysfunction, and survival rate in LPS-treated rats.