Learn More
Medchemexpress LLC Carboxy-PTIO potassium | 148819-94-7 | 99.0% | 315.39 | 50 MG
SDP
Catalog No. 5000388023
Encompass_Preferred
Carboxy-PTIO potassium is a potent nitric oxide (NO) scavenger that rapidly reacts with NO to produce NO2. It can prevent hypotension and endotoxic shock through its direct NO-scavenging action in a lipopolysaccharide-stimulated rat model. This compound is intended for research use only and is not sold to patients.
  • Potent nitric oxide (NO) scavenger
  • Reacts quickly with NO to produce NO2
  • Can prevent hypotension and endotoxic shock by scavenging NO in LPS-stimulated rat models
  • Significantly suppresses stimulation of NO expression induced by Physalin A treatment in vitro
  • Reduces Physalin A-induced cleavage of procaspase-3 and PARP, and down-regulates ICAD expression, diminishing DNA fragmentation in nuclei
  • Reverses decreased mTOR and p-mTOR levels induced by Physalin A, with concomitant suppression of LC3 I to LC3 II conversions in A375-S2 cells
Catalog No. 50-003-88023 Supplier Medchemexpress LLC Supplier No. HY18734A50MG
Carboxy-PTIO potassium is a potent nitric oxide (NO) scavenger that rapidly reacts with NO to produce NO2. It can prevent hypotension and endotoxic shock through its direct NO-scavenging action in a lipopolysaccharide-stimulated rat model. This compound is intended for research use only and is not sold to patients.
  • Potent nitric oxide (NO) scavenger
  • Reacts quickly with NO to produce NO2
  • Can prevent hypotension and endotoxic shock by scavenging NO in LPS-stimulated rat models
  • Significantly suppresses stimulation of NO expression induced by Physalin A treatment in vitro
  • Reduces Physalin A-induced cleavage of procaspase-3 and PARP, and down-regulates ICAD expression, diminishing DNA fragmentation in nuclei
  • Reverses decreased mTOR and p-mTOR levels induced by Physalin A, with concomitant suppression of LC3 I to LC3 II conversions in A375-S2 cells