Learn More
Medchemexpress LLC HY-112680A 1mg Medchemexpress, Carboxy pyridostatin (trifluoroacetate salt) CAS:2444713-88-4 Purity:>98%
SDP
Catalog No. 502029734
Encompass_Preferred
Medchemexpress, HY-112680A 1mg Carboxy pyridostatin (trifluoroacetate salt) CAS:2444713-88-4 Carboxy pyridostatin trifluoroacetate salt has the peculiarity to exhibit high molecular specificity for RNA over DNA G4s. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9734 Supplier Medchemexpress LLC Supplier No. HY112680A1MG
Medchemexpress, HY-112680A 1mg Carboxy pyridostatin (trifluoroacetate salt) CAS:2444713-88-4 Carboxy pyridostatin trifluoroacetate salt has the peculiarity to exhibit high molecular specificity for RNA over DNA G4s. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.