Learn More
Medchemexpress LLC Carboxy pyridostatin trifluoroacetate salt | 2444713-88-4 | 99.5% | 820.73 | 100 MG
SDP
Catalog No. 5000436995
Encompass_Preferred
Carboxy pyridostatin trifluoroacetate salt is a G-quadruplex ligand with high molecular specificity to RNA on DNA G4s, and it also reduces ATF-5 protein. This compound is known to decrease cell proliferation and inhibit stress granule (SG) formation. It is intended for research use only.
  • Reduces the size of adult SVZ-derived neurospheres and the number of cells in neurospheres.
  • Blocks proliferation and neurosphere fusion in adult SVZ-derived neurosphere cultures.
  • Hinders stress granule (SG) formation in U2OS cells.
  • Significantly reduces the number of PCNA-expressing cells in the adult SVZ.
  • Increases the number of OLIG2-expressing cells in the corpus callosum in C57BL/6 mice.
  • Affects the differentiation of neural stem and progenitor cells in different brain regions.
Catalog No. 50-004-36995 Supplier Medchemexpress LLC Supplier No. HY112680A100MG
Carboxy pyridostatin trifluoroacetate salt is a G-quadruplex ligand with high molecular specificity to RNA on DNA G4s, and it also reduces ATF-5 protein. This compound is known to decrease cell proliferation and inhibit stress granule (SG) formation. It is intended for research use only.
  • Reduces the size of adult SVZ-derived neurospheres and the number of cells in neurospheres.
  • Blocks proliferation and neurosphere fusion in adult SVZ-derived neurosphere cultures.
  • Hinders stress granule (SG) formation in U2OS cells.
  • Significantly reduces the number of PCNA-expressing cells in the adult SVZ.
  • Increases the number of OLIG2-expressing cells in the corpus callosum in C57BL/6 mice.
  • Affects the differentiation of neural stem and progenitor cells in different brain regions.