Learn More
Medchemexpress LLC Carboxy pyridostatin trifluoroacetate salt | 2444713-88-4 | 99.5% | 820.73 | 25 MG
SDP
Catalog No. 5000439579
Encompass_Preferred
Carboxy pyridostatin trifluoroacetate salt is a G-quadruplex ligand. It exhibits high molecular specificity to RNA on DNA G4s and reduces ATF-5 protein. Additionally, it reduces cell proliferation and hinders stress granule (SG) formation.
  • G-quadruplex ligand
  • High molecular specificity to RNA on DNA G4s
  • Reduces ATF-5 protein
  • Reduces cell proliferation
  • Hinders stress granule (SG) formation
  • Reduces the size of adult SVZ-derived neurospheres and the number of cells in neurospheres
  • Blocks proliferation and neurosphere fusion in adult SVZ-derived neurosphere cultures
  • Significantly reduces the number of PCNA-expressing cells in the adult SVZ in mice
  • Increases the number of OLIG2-expressing cells in the corpus callosum in mice
Catalog No. 50-004-39579 Supplier Medchemexpress LLC Supplier No. 25MGHY112680A
Carboxy pyridostatin trifluoroacetate salt is a G-quadruplex ligand. It exhibits high molecular specificity to RNA on DNA G4s and reduces ATF-5 protein. Additionally, it reduces cell proliferation and hinders stress granule (SG) formation.
  • G-quadruplex ligand
  • High molecular specificity to RNA on DNA G4s
  • Reduces ATF-5 protein
  • Reduces cell proliferation
  • Hinders stress granule (SG) formation
  • Reduces the size of adult SVZ-derived neurospheres and the number of cells in neurospheres
  • Blocks proliferation and neurosphere fusion in adult SVZ-derived neurosphere cultures
  • Significantly reduces the number of PCNA-expressing cells in the adult SVZ in mice
  • Increases the number of OLIG2-expressing cells in the corpus callosum in mice