Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carboxy pyridostatin trifluoroacetate salt | 2444713-88-4 | 99.5% | 820.73 g/mol | C37H35F3N10O9 | 5 MG
SDP

Catalog No. 5000262464
Encompass_Preferred

Carboxy pyridostatin (trifluoroacetate salt) is a research-grade G-quadruplex ligand supplied as a solid salt for in vitro and in vivo studies. It selectively recognizes RNA on DNA G-quadruplex structures, reduces ATF-5 protein levels, inhibits cell proliferation, and impairs stress granule formation.

  • Cas number: 2444713-88-4.
  • Molecular formula: C37H35F3N10O9.
  • Molecular weight: 820.73 g/mol.
  • Purity: 99.5%.
  • Pack sizes include 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
  • Soluble in DMSO at 50 mg/mL with ultrasonic warming to 60°C.
  • In vivo formulations achievable at ≥2.5 mg/mL in common vehicle mixtures.
  • Storage: solid sealed away from moisture, recommended -20°C; in solution -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-62464 Supplier Medchemexpress LLC Supplier No. HY112680A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carboxy pyridostatin (trifluoroacetate salt) is a research-grade G-quadruplex ligand supplied as a solid salt for in vitro and in vivo studies. It selectively recognizes RNA on DNA G-quadruplex structures, reduces ATF-5 protein levels, inhibits cell proliferation, and impairs stress granule formation.

  • Cas number: 2444713-88-4.
  • Molecular formula: C37H35F3N10O9.
  • Molecular weight: 820.73 g/mol.
  • Purity: 99.5%.
  • Pack sizes include 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
  • Soluble in DMSO at 50 mg/mL with ultrasonic warming to 60°C.
  • In vivo formulations achievable at ≥2.5 mg/mL in common vehicle mixtures.
  • Storage: solid sealed away from moisture, recommended -20°C; in solution -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.