Learn More
Medchemexpress LLC Carboxyamidotriazole Orotate | 187739-60-2 | 99.7% | 580.76 | 5 MG
SDP
Catalog No. 5000392590
Encompass_Preferred
Carboxyamidotriazole Orotate is the orotate salt form of Carboxyamidotriazole (CAI), an orally bioavailable signal transduction inhibitor. It functions as a cytostatic inhibitor of nonvoltage-operated calcium channels and calcium channel-mediated signaling pathways. This product demonstrates anti-tumor, anti-inflammatory, and antiangiogenic effects.
  • Functions as a cytostatic inhibitor of nonvoltage-operated calcium channels.
  • Inhibits calcium channel-mediated signaling pathways.
  • Exhibits anti-tumor effects.
  • Exhibits anti-inflammatory effects.
  • Exhibits antiangiogenic effects.
  • Inhibits cell proliferation of LAMA84R and K562R cell lines.
  • Reduces phosphorylation of CrkL.
  • Shows dose-dependent inhibition of total and phosphorylated Bcr-Abl levels.
  • Displays antitumor activity on CML xenografts.
  • Increased survival in animal models.
Catalog No. 50-003-92590 Supplier Medchemexpress LLC Supplier No. HY161255MG
Carboxyamidotriazole Orotate is the orotate salt form of Carboxyamidotriazole (CAI), an orally bioavailable signal transduction inhibitor. It functions as a cytostatic inhibitor of nonvoltage-operated calcium channels and calcium channel-mediated signaling pathways. This product demonstrates anti-tumor, anti-inflammatory, and antiangiogenic effects.
  • Functions as a cytostatic inhibitor of nonvoltage-operated calcium channels.
  • Inhibits calcium channel-mediated signaling pathways.
  • Exhibits anti-tumor effects.
  • Exhibits anti-inflammatory effects.
  • Exhibits antiangiogenic effects.
  • Inhibits cell proliferation of LAMA84R and K562R cell lines.
  • Reduces phosphorylation of CrkL.
  • Shows dose-dependent inhibition of total and phosphorylated Bcr-Abl levels.
  • Displays antitumor activity on CML xenografts.
  • Increased survival in animal models.