Learn More
Medchemexpress LLC Carboxyamidotriazole orotate | 187739-60-2 | 99.7% | 580.76 | 50 MG
SDP
Catalog No. 5000393066
Encompass_Preferred
Carboxyamidotriazole Orotate is an orally bioavailable signal transduction inhibitor. It acts as a cytostatic inhibitor of nonvoltage-operated calcium channels and calcium channel-mediated signaling pathways, demonstrating anti-tumor, anti-inflammatory, and antiangiogenic effects.
  • Inhibits cell proliferation in specific leukemia cell lines.
  • Reduces phosphorylation of Bcr-Abl kinase targets.
  • Induces a 50% growth reduction in chronic myelogenous leukemia (CML) cell lines.
  • Demonstrates dose-dependent inhibition of Bcr-Abl levels.
  • Exhibits antitumor activity in CML xenografts, leading to increased survival.
  • Has been evaluated in multiple clinical trials for various cancers.
  • Intended for research use only.
Catalog No. 50-003-93066 Supplier Medchemexpress LLC Supplier No. HY1612550MG
Carboxyamidotriazole Orotate is an orally bioavailable signal transduction inhibitor. It acts as a cytostatic inhibitor of nonvoltage-operated calcium channels and calcium channel-mediated signaling pathways, demonstrating anti-tumor, anti-inflammatory, and antiangiogenic effects.
  • Inhibits cell proliferation in specific leukemia cell lines.
  • Reduces phosphorylation of Bcr-Abl kinase targets.
  • Induces a 50% growth reduction in chronic myelogenous leukemia (CML) cell lines.
  • Demonstrates dose-dependent inhibition of Bcr-Abl levels.
  • Exhibits antitumor activity in CML xenografts, leading to increased survival.
  • Has been evaluated in multiple clinical trials for various cancers.
  • Intended for research use only.