Learn More
Medchemexpress LLC Carboxyamidotriazole (orotate) | 187739-60-2 | MFCD22572756 | 99.7% | 580.76 g·mol⁻¹ | C22H16Cl3N7O6 | 10 MG
SDP
Catalog No. 5000288560
Encompass_Preferred
Carboxyamidotriazole orotate is the orotate salt of carboxyamidotriazole, an orally active calcium influx inhibitor used in research on calcium-mediated signaling, angiogenesis, inflammation, and tumor biology. It is supplied as a solid intended for laboratory research.
  • Acts as a calcium influx inhibitor with anti-tumor and anti-inflammatory activity.
  • High purity (~99.7%) suitable for research applications.
  • Solid, off-white to yellow appearance for stable handling and storage.
  • Molecular weight 580.76 and CAS number 187739-60-2 for precise identification.
  • Store sealed and away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Available in milligram quantities for preclinical and in vitro studies.
Catalog No. 50-002-88560 Supplier Medchemexpress LLC Supplier No. HY1612510MG
Carboxyamidotriazole orotate is the orotate salt of carboxyamidotriazole, an orally active calcium influx inhibitor used in research on calcium-mediated signaling, angiogenesis, inflammation, and tumor biology. It is supplied as a solid intended for laboratory research.
  • Acts as a calcium influx inhibitor with anti-tumor and anti-inflammatory activity.
  • High purity (~99.7%) suitable for research applications.
  • Solid, off-white to yellow appearance for stable handling and storage.
  • Molecular weight 580.76 and CAS number 187739-60-2 for precise identification.
  • Store sealed and away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Available in milligram quantities for preclinical and in vitro studies.