Learn More
Selleck Chemical LLC CarboxyaMidotriazole orotate S2975-5mg
SDP
Catalog No. 503109619
Encompass_Preferred
Carboxyamidotriazole Orotate (L-651582 Orotate) is the orotate salt form of Carboxyamidotriazole (CAI) an orally bioavailable signal transduction inhibitor Carboxyamidotriazole Orotate is a cytostatic inhibitor of nonvoltage-operated calcium channels and calcium channel-mediated signaling pathways Carboxyamidotriazole Orotate inhibits angiogenesis tumor growth invasion and metastasis
Catalog No. 50-310-9619 Supplier Selleck Chemical LLC Supplier No. S29755MG
Carboxyamidotriazole Orotate (L-651582 Orotate) is the orotate salt form of Carboxyamidotriazole (CAI) an orally bioavailable signal transduction inhibitor Carboxyamidotriazole Orotate is a cytostatic inhibitor of nonvoltage-operated calcium channels and calcium channel-mediated signaling pathways Carboxyamidotriazole Orotate inhibits angiogenesis tumor growth invasion and metastasis