Learn More
TARGETMOL CHEMICALS INC CARBOXYATRACTYLOSIDE 25MG
SDP
Catalog No. 502579100
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Carboxyatractyloside (Carboxyatractyloside tripotassium salt) is a natural product. It acts as an inhibitor of ADP/ATP carrier inhibits mitochondrial ADP/ATP transport. purity: 99%
Catalog No. 50-257-9100 Supplier TARGETMOL CHEMICALS INC Supplier No. T817025MG
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Carboxyatractyloside (Carboxyatractyloside tripotassium salt) is a natural product. It acts as an inhibitor of ADP/ATP carrier inhibits mitochondrial ADP/ATP transport. purity: 99%