Learn More
Medchemexpress LLC HY-N2522 5mg Medchemexpress, Carboxyatractyloside (dipotassium) CAS:33286-30-5 Purity:>98%
SDP
Catalog No. 501970401
Encompass_Preferred
Medchemexpress, HY-N2522 5mg Carboxyatractyloside (dipotassium) CAS:33286-30-5 Carboxyatractyloside dipotassium is a toxic natural product, acts as an inhibitor of ADP/ATP carrier, inhibits mitochondrial ADP/ATP transport. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0401 Supplier Medchemexpress LLC Supplier No. HYN25225MG
Medchemexpress, HY-N2522 5mg Carboxyatractyloside (dipotassium) CAS:33286-30-5 Carboxyatractyloside dipotassium is a toxic natural product, acts as an inhibitor of ADP/ATP carrier, inhibits mitochondrial ADP/ATP transport. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.