Learn More
Medchemexpress LLC Carboxyatractyloside tripotassium | 77228-71-8 | 885.09 | 1 MG
SDP
Catalog No. 5000415461
Encompass_Preferred
Carboxyatractyloside tripotassium is a diterpenoid that can be isolated from plants of the genus Xanthium. It acts as an ADP/ATP carrier inhibitor, specifically inhibiting mitochondrial ADP/ATP transport. This compound promotes ROS production, induces Ca2+ release, and leads to mitochondrial dysfunction. In rats, Carboxyatractyloside tripotassium has been observed to induce lethargy, weakness, and epileptic seizures.
  • Inhibits mitochondrial ADP/ATP transport.
  • Promotes production of reactive oxygen species (ROS).
  • Induces Ca2+ release.
  • Leads to mitochondrial dysfunction.
  • Can be isolated from Xanthium plants.
Catalog No. 50-004-15461 Supplier Medchemexpress LLC Supplier No. HYN15021MG
Carboxyatractyloside tripotassium is a diterpenoid that can be isolated from plants of the genus Xanthium. It acts as an ADP/ATP carrier inhibitor, specifically inhibiting mitochondrial ADP/ATP transport. This compound promotes ROS production, induces Ca2+ release, and leads to mitochondrial dysfunction. In rats, Carboxyatractyloside tripotassium has been observed to induce lethargy, weakness, and epileptic seizures.
  • Inhibits mitochondrial ADP/ATP transport.
  • Promotes production of reactive oxygen species (ROS).
  • Induces Ca2+ release.
  • Leads to mitochondrial dysfunction.
  • Can be isolated from Xanthium plants.