Learn More
Medchemexpress LLC Carboxyatractyloside tripotassium | 77228-71-8 | 885.09 | 5 MG
SDP
Catalog No. 5000414747
Encompass_Preferred
Carboxyatractyloside tripotassium is a diterpenoid that can be isolated from plants of the genus Xanthium. It functions as an ADP/ATP carrier inhibitor, specifically inhibiting mitochondrial ADP/ATP transport. This compound promotes ROS production, induces Ca2+ release, and leads to mitochondrial dysfunction, and has been observed to induce lethargy, weakness, and epileptic seizures in rats.
  • Promotes cell death in primary mouse lung endothelial cells.
  • Exacerbates mitochondrial dysfunction, including depolarization, increased ROS generation, and MPT pore opening.
  • Induces Ca2+ release, mitochondrial swelling, and membrane potential collapse in aged rat liver mitochondria.
  • Causes histopathological damage to the liver and kidneys, including vacuolar degeneration and necrosis of hepatocytes and renal tubular necrosis.
Catalog No. 50-004-14747 Supplier Medchemexpress LLC Supplier No. HYN15025MG
Carboxyatractyloside tripotassium is a diterpenoid that can be isolated from plants of the genus Xanthium. It functions as an ADP/ATP carrier inhibitor, specifically inhibiting mitochondrial ADP/ATP transport. This compound promotes ROS production, induces Ca2+ release, and leads to mitochondrial dysfunction, and has been observed to induce lethargy, weakness, and epileptic seizures in rats.
  • Promotes cell death in primary mouse lung endothelial cells.
  • Exacerbates mitochondrial dysfunction, including depolarization, increased ROS generation, and MPT pore opening.
  • Induces Ca2+ release, mitochondrial swelling, and membrane potential collapse in aged rat liver mitochondria.
  • Causes histopathological damage to the liver and kidneys, including vacuolar degeneration and necrosis of hepatocytes and renal tubular necrosis.