Learn More
Medchemexpress LLC Carboxybenzoyl-leucyl-arginine 7-amido-4-methylcoumarin | 156192-32-4 | 99.2% | 578.66 g/mol | C30H38N6O6 | 5 MG
SDP
Catalog No. 5000221596
Encompass_Preferred
Z-Leu-Arg-AMC is a fluorogenic peptide substrate for assaying cysteine proteases such as cathepsin K. It consists of a carboxybenzoyl (Z)-protected leucyl-arginine linked to 7-amido-4-methylcoumarin (AMC); enzymatic cleavage releases fluorescent AMC for sensitive kinetic measurements. Supplied as a solid or as a 10 mM solution in DMSO for research use only.
  • Sensitive fluorogenic reporter released on protease-mediated cleavage.
  • Suitable for cathepsin K and other cysteine protease activity assays.
  • High reported purity for consistent, reproducible results.
  • Available as solid or pre-prepared DMSO solution for flexible use.
  • Soluble in DMSO; handle and store according to standard reagent recommendations.
Catalog No. 50-002-21596 Supplier Medchemexpress LLC Supplier No. HY1420215MG
Z-Leu-Arg-AMC is a fluorogenic peptide substrate for assaying cysteine proteases such as cathepsin K. It consists of a carboxybenzoyl (Z)-protected leucyl-arginine linked to 7-amido-4-methylcoumarin (AMC); enzymatic cleavage releases fluorescent AMC for sensitive kinetic measurements. Supplied as a solid or as a 10 mM solution in DMSO for research use only.
  • Sensitive fluorogenic reporter released on protease-mediated cleavage.
  • Suitable for cathepsin K and other cysteine protease activity assays.
  • High reported purity for consistent, reproducible results.
  • Available as solid or pre-prepared DMSO solution for flexible use.
  • Soluble in DMSO; handle and store according to standard reagent recommendations.