Learn More
Medchemexpress LLC Carboxybetaine metha 10g
SDP
Catalog No. 5000204933
Encompass_Preferred
Carboxybetaine methacrylate is an amphoteric ionic compound with excellent properties such as good hydrophilicity antistatic property and biocompatibility Carboxybetaine methacrylate can be used to prepare tissue engineering scaffolds drug carriers and so on[1][2]
Catalog No. 50-002-04933 Supplier Medchemexpress LLC Supplier No. HYW11058410G
Carboxybetaine methacrylate is an amphoteric ionic compound with excellent properties such as good hydrophilicity antistatic property and biocompatibility Carboxybetaine methacrylate can be used to prepare tissue engineering scaffolds drug carriers and so on[1][2]