Learn More
Medchemexpress LLC Carboxybetaine metha 250mg
SDP
Catalog No. 5000206018
Encompass_Preferred
Carboxybetaine methacrylate is an amphoteric ionic compound with excellent properties such as good hydrophilicity antistatic property and biocompatibility Carboxybetaine methacrylate can be used to prepare tissue engineering scaffolds drug carriers and so on[1][2]
Catalog No. 50-002-06018 Supplier Medchemexpress LLC Supplier No. 250MGHYW110584
Carboxybetaine methacrylate is an amphoteric ionic compound with excellent properties such as good hydrophilicity antistatic property and biocompatibility Carboxybetaine methacrylate can be used to prepare tissue engineering scaffolds drug carriers and so on[1][2]