Learn More
Medchemexpress LLC Carboxybetaine metha 5g
SDP
Catalog No. 5000206738
Encompass_Preferred
Carboxybetaine methacrylate is an amphoteric ionic compound with excellent properties such as good hydrophilicity antistatic property and biocompatibility Carboxybetaine methacrylate can be used to prepare tissue engineering scaffolds drug carriers and so on[1][2]
Catalog No. 50-002-06738 Supplier Medchemexpress LLC Supplier No. HYW1105845G
Carboxybetaine methacrylate is an amphoteric ionic compound with excellent properties such as good hydrophilicity antistatic property and biocompatibility Carboxybetaine methacrylate can be used to prepare tissue engineering scaffolds drug carriers and so on[1][2]