Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carboxyethyl-γ-aminobutyric acid | 4386-03-2 | MFCD00057954 | 98.0% | 175.18 g/mol | C7H13NO4 | 100 MG
SDP

Catalog No. 5000284415
Encompass_Preferred

Carboxyethyl-γ-aminobutyric acid is a small-molecule biochemical assay reagent intended for laboratory research and assay applications. It is supplied as a characterized solid with supporting datasheet and certificate of analysis for identity and purity.

  • Used as a biochemical assay reagent
  • Purity 98.0%
  • Molecular weight 175.18 g/mol
  • Chemical formula C7H13NO4
  • CAS number 4386-03-2
  • Physical form: white to off-white solid powder
  • Storage: powder -20°C for long-term, 4°C for short-term
  • Available quantity: 100 mg

Catalog No. 50-002-84415 Supplier Medchemexpress LLC Supplier No. HYW593287100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carboxyethyl-γ-aminobutyric acid is a small-molecule biochemical assay reagent intended for laboratory research and assay applications. It is supplied as a characterized solid with supporting datasheet and certificate of analysis for identity and purity.

  • Used as a biochemical assay reagent
  • Purity 98.0%
  • Molecular weight 175.18 g/mol
  • Chemical formula C7H13NO4
  • CAS number 4386-03-2
  • Physical form: white to off-white solid powder
  • Storage: powder -20°C for long-term, 4°C for short-term
  • Available quantity: 100 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.