Learn More
BIOHIPPO Carboxyethyl-Cysteine mAb
SDP
Catalog No. 5000975485
Encompass
U-Blot Pan-Carboxyethyl-Cysteine Rabbit mAb is a Rabbit-derived monoclonal antibody that specifically recognizes Carboxyethyl-Cysteine a cysteine carboxyethylation modification at Cys(C) Carboxyethyl-Cysteine (CEC) is a novel post-translational modification occurring on cysteine residues of proteins This modification is formed by covalently attaching a carboxyethyl group to the thiol group (-SH) of cysteine generating a stable thioether linkage CEC modification is widely distributed on histones and various functional proteins and has been reported to be involved in cellular oxidative stress metabolic regulation protein stability and a variety of physiological and pathological processes making it an important target in protein modification and disease research Supplied as 100 uL
Catalog No. 50-009-75485 Supplier BIOHIPPO Supplier No. BHA21821316
U-Blot Pan-Carboxyethyl-Cysteine Rabbit mAb is a Rabbit-derived monoclonal antibody that specifically recognizes Carboxyethyl-Cysteine a cysteine carboxyethylation modification at Cys(C) Carboxyethyl-Cysteine (CEC) is a novel post-translational modification occurring on cysteine residues of proteins This modification is formed by covalently attaching a carboxyethyl group to the thiol group (-SH) of cysteine generating a stable thioether linkage CEC modification is widely distributed on histones and various functional proteins and has been reported to be involved in cellular oxidative stress metabolic regulation protein stability and a variety of physiological and pathological processes making it an important target in protein modification and disease research Supplied as 100 uL